Dinatriumhexachloropalladat ≥98%
Lieferant: ABCR GMBH
|
Achtung
|
Synonyme:
Natriumhexachloropalladat(IV)
ABCRAB121388.0001EA
0
EUR
ABCRAB121388.0001
Dinatriumhexachloropalladat ≥98%
Dinatriumhexachloropalladat
Formel:
Cl₆Na₂Pd Molecular Weight: 365,12 g/mol |
MDL:
MFCD00014235 CAS-Nummer: 53823-60-2 EINECS: 258-809-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...