Dibenzosuberylchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Chlordibenzo[a,d]cycloheptan5-Chlor-10,11-dihydro-5H-dibenzo[a,d]cyclohepten

M-Clarity™ quality level = MQ100

SIALC34308-5GEA 47.4 EUR
SIALC34308-5G
Dibenzosuberylchlorid, Sigma-Aldrich®
Dibenzosuberylchlorid
Formel: C₁₅H₁₃Cl
Molecular Weight: 228,72 g/mol
Schmelzpunkt: 108 °C
MDL: MFCD00003592
CAS-Nummer: 1210-33-9
EINECS: 214-910-2
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR