3,6-Dibromchromon

Lieferant: CHEMPUR

CHMPFL12058.POREA 0 EUR
CHMPFL12058.POR
3,6-Dibromchromon
3,6-Dibromchromon
Formel: C₉H₄Br₂O₂
CAS-Nummer: 115237-39-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR