Monomethylphthalat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ200

SIAL317640-25GEA 71.2 EUR
SIAL317640-25G
Monomethylphthalat, Sigma-Aldrich®
Monomethylphthalat
Formel: C₉H₈O₄
Molecular Weight: 180,16 g/mol
MDL: MFCD00002466
CAS-Nummer: 4376-18-5
EINECS: 224-476-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR