Monomethylphthalat

Lieferant: EHRENSTORFER GMBH

EHERC16179000EA 40 EUR
EHERC16179000
Monomethylphthalat
Monomethylphthalat
Formel: C₉H₈O₄
Molecular Weight: 180,16
MDL: MFCD00002466
CAS-Nummer: 4376-18-5
EINECS: 224-476-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR