3-Brom-2-methyl-5-(pentafluorthio)anilin ≥97%
|
Gefahr
|
H33765.MDEA
128
EUR
H33765.MD
3-Brom-2-methyl-5-(pentafluorthio)anilin ≥97%
3-Brom-2-methyl-5-(pentafluorthio)anilin
Formel:
C₇H₇BrF₅NS Molecular Weight: 312,10 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22201077 CAS-Nummer: 1373920-91-2 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...