4-(1-Piperidinyl)anilin, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL556629-1GEA
45.3
EUR
SIAL556629-1G
4-(1-Piperidinyl)anilin, Sigma-Aldrich®
4-(1-Piperidinyl)anilin
Formel:
C₁₁H₁₆N₂ Molecular Weight: 176,26 g/mol |
MDL:
MFCD00051688 CAS-Nummer: 2359-60-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...