(R)-(-)-1-Benzyl-3-hydroxypiperidin, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL455172-5GEA
229
EUR
SIAL455172-5G
(R)-(-)-1-Benzyl-3-hydroxypiperidin, Sigma-Aldrich®
(R)-(-)-1-Benzyl-3-hydroxypiperidin
Formel:
C₁₂H₁₇NO Molecular Weight: 191,27 g/mol |
MDL:
MFCD00274307 CAS-Nummer: 91599-81-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...