2-Amino-N-sec-butylbenzamid ≥96%
|
Achtung
|
H50724.03EA
91.4
EUR
H50724.03
2-Amino-N-sec-butylbenzamid ≥96%
2-Amino-N-sec-butylbenzamid
Formel:
C₁₁H₁₆N₂O Molecular Weight: 192,26 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00116303 CAS-Nummer: 30391-87-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...