3-Chlor-2,5-dimethylpyrazin, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIALC38400-25GEA
65.3
EUR
SIALC38400-25G
3-Chlor-2,5-dimethylpyrazin, Sigma-Aldrich®
3-Chlor-2,5-dimethylpyrazin
Formel:
C₆H₇ClN₂ Molecular Weight: 142,5900 g/mol |
MDL:
MFCD00006143 CAS-Nummer: 95-89-6 EINECS: 202-463-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...