2-Methoxy-4-nitrobenzonitril, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL524751-5GEA
34.5
EUR
SIAL524751-5G
2-Methoxy-4-nitrobenzonitril, Sigma-Aldrich®
2-Methoxy-4-nitrobenzonitril
Formel:
C₈H₆N₂O₃ Molecular Weight: 178,15 g/mol |
MDL:
MFCD02093781 CAS-Nummer: 101084-96-2 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...