5-Brom-4-cyclopentylpyrimidin
Lieferant: COMBI-BLOCKS INC.
COBBHC-6074-5GEA
541
EUR
COBBHC-6074-5G
COBBHC-6074-1G
5-Brom-4-cyclopentylpyrimidin
5-Bromo-4-cyclopentylpyrimidine
Formel:
C₉H₁₁BrN₂ |
MDL:
MFCD09800933 CAS-Nummer: 951884-32-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...