5-Brom-4-octylpyrimidin

Lieferant: COMBI-BLOCKS INC.

COBBHC-6115-1GEA 89.5 EUR
COBBHC-6115-1G COBBHC-6115-5G
5-Brom-4-octylpyrimidin
5-Bromo-4-octylpyrimidine
Formel: C₁₂H₁₉BrN₂
MDL: MFCD10699650
CAS-Nummer: 1033201-80-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR