5-Brom-4-sec-butylpyrimidin
Lieferant: COMBI-BLOCKS INC.
COBBHC-6073-5GEA
541
EUR
COBBHC-6073-5G
COBBHC-6073-1G
5-Brom-4-sec-butylpyrimidin
5-Brom-4-sec-butylpyrimidin
Formel:
C₈H₁₁BrN₂ |
MDL:
MFCD09800932 CAS-Nummer: 951884-30-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...