4-Isopropylphenylhydrazin Hydrochlorid

Lieferant: CHEMPUR

CHMPFL13616.POREA 0 EUR
CHMPFL13616.POR
4-Isopropylphenylhydrazin Hydrochlorid
4-Isopropylphenylhydrazin Hydrochlorid
Formel: C₉H₁₅ClN₂
Molecular Weight: 186,69 g/mol
MDL: MFCD00013461
CAS-Nummer: 118427-29-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR