4-Nitro-1H-pyrazol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL696161-5GEA 55 EUR
SIAL696161-5G
4-Nitro-1H-pyrazol, Sigma-Aldrich®
4-Nitro-1H-pyrazole
Formel: C₃H₃N₃O₂
Molecular Weight: 113,0800 g/mol
MDL: MFCD00159626
CAS-Nummer: 2075-46-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR