4-Nitro-1H-pyrazol
Lieferant: COMBI-BLOCKS INC.
COBBOR-0595-25GEA
36.9
EUR
COBBOR-0595-25G
4-Nitro-1H-pyrazol
4-Nitro-1H-pyrazole
Formel:
C₃H₃N₃O₂ Molecular Weight: 113,0800 g/mol |
MDL:
MFCD00159626 CAS-Nummer: 2075-46-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...