Zirkonium(IV)sulfat Hydrat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL366773-25GEA 396 EUR
SIAL366773-25G SIAL366773-125G
Zirkonium(IV)sulfat Hydrat, Sigma-Aldrich®
Zirkonium(IV)sulfat Hydrat
Formel: O₈S₂Zr·H₂O
MDL: MFCD00167043
CAS-Nummer: 34806-73-0
EINECS: 238-694-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR