Ethyl-2-methylnicotinat, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL325201-5GEA
77.4
EUR
SIAL325201-5G
Ethyl-2-methylnicotinat, Sigma-Aldrich®
Ethyl-2-methylnicotinat
Formel:
C₉H₁₁NO₂ Molecular Weight: 165,19 g/mol |
MDL:
MFCD00006336 CAS-Nummer: 1721-26-2 EINECS: 217-013-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...