Methyl-6-fluorsalicylat

Lieferant: Apollo Scientific

APOSPC5092-1GEA 55.1 EUR
APOSPC5092-1G
Methyl-6-fluorsalicylat
Methyl-6-fluorsalicylat
Formel: C₈H₇FO₃
MDL: MFCD00153207
CAS-Nummer: 72373-81-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR