1-Brom-3,5-di-tert-butylbenzol

Lieferant: COMBI-BLOCKS INC.

COBBOS-1034-100GEA 331 EUR
COBBOS-1034-100G
1-Brom-3,5-di-tert-butylbenzol
1-Brom-3,5-di-tert-butylbenzol
Formel: C₁₄H₂₁Br
Molecular Weight: 269,23 g/mol
MDL: MFCD00796945
CAS-Nummer: 22385-77-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR