3-Fluoro-4-methylanisole, 97%

H32472.06EA 141 EUR
ALFAH32472.06 ALFAH32472.03
3-Fluoro-4-methylanisole, 97%
Purity: 97%
CAS No.: 405-06-1
Molecular Formula: C8H9FO
Formula Weight: 140.16
Refractive Index: 1.4909
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR