3-Nitrosalicylsäure, Sigma-Aldrich®

Lieferant: Merck

SIAL73815-5GEA 124 EUR
SIAL73815-5G
3-Nitrosalicylsäure, Sigma-Aldrich®
3-Nitrosalicylsäure
Formel: C₇H₅NO₅
Molecular Weight: 183,1183 g/mol
MDL: MFCD00024240
CAS-Nummer: 85-38-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR