4-Brom-2-ethoxyanisol
Lieferant: COMBI-BLOCKS INC.
COBBOT-0640-1GEA
121
EUR
COBBOT-0640-1G
COBBOT-0640-5G
4-Brom-2-ethoxyanisol
4-Brom-2-ethoxyanisol
Formel:
C₉H₁₁BrO₂ |
MDL:
MFCD09972175 CAS-Nummer: 52849-52-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...