3-Bromcarbazol

MAYBJFD01173CBEA 0 EUR
MAYBJFD01173CB MAYBJFD01173DA
3-Bromcarbazol
3-Bromocarbazole
Formel: C₁₂H₈BrN
Molecular Weight: 246,11
MDL: MFCD00222621
CAS-Nummer: 1592-95-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR