Ethylphenylcyanacetat

Lieferant: CHEMPUR

Synonyme: Ethyl 2-cyano-2-phenylacetate

CHMPFL13578.POREA 0 EUR
CHMPFL13578.POR
Ethylphenylcyanacetat
Ethylphenylcyanacetat
Formel: C₁₁H₁₁NO₂
Molecular Weight: 189,21 g/mol
MDL: MFCD00001866
CAS-Nummer: 4553-07-5
EINECS: 224-921-4
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR