4-Methoxy-N-methylanilin, Sigma-Aldrich®

Lieferant: Merck

SIAL180033-10GEA 230 EUR
SIAL180033-10G SIAL180033-1G
4-Methoxy-N-methylanilin, Sigma-Aldrich®
4-Methoxy-N-methylanilin
Formel: C₈H₁₁NO
Molecular Weight: 137,18 g/mol
MDL: MFCD00008399
CAS-Nummer: 5961-59-1
EINECS: 227-735-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR