3-Isopropoxypropylamin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL550000-100MLEA 32.9 EUR
SIAL550000-100ML SIAL550000-500ML
3-Isopropoxypropylamin, Sigma-Aldrich®
3-Isopropoxypropylamin
Formel: C₆H₁₅NO
Molecular Weight: 117,19
MDL: MFCD00008220
CAS-Nummer: 2906-12-9
EINECS: 220-816-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR