Hafniumdichloridoxid Hydrat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL229652-1GEA 189 EUR
SIAL229652-1G
Hafniumdichloridoxid Hydrat, Sigma-Aldrich®
Hafniumdichloridoxid Hydrat
Formel: Cl₂HfO·H₂O
MDL: MFCD00150415
CAS-Nummer: 15461-28-6
EINECS: 237-345-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR