5-Brom-4,6-difluorpyrimidin, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL700975-250MGEA
140
EUR
SIAL700975-250MG
5-Brom-4,6-difluorpyrimidin, Sigma-Aldrich®
5-Bromo-4,6-difluoropyrimidine
Formel:
C₄HBrF₂N₂ Molecular Weight: 194,9650 g/mol |
MDL:
MFCD08686978 CAS-Nummer: 946681-88-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...