2,6-Dichlor-4-nitropyridin-N-oxid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL673161-5GEA
93.7
EUR
SIAL673161-5G
2,6-Dichlor-4-nitropyridin-N-oxid, Sigma-Aldrich®
2,6-Dichlor-4-nitropyridin-N-oxid
Formel:
C₅H₂Cl₂N₂O₃ Molecular Weight: 208,99 g/mol |
MDL:
MFCD00955611 CAS-Nummer: 2587-01-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...