Zirkoniumcarbid

Lieferant: ABCR GMBH

ABCRAB134581.1000EA 0 EUR
ABCRAB134581.1000
Zirkoniumcarbid
Zirconium carbide
Formel: CZr
Molecular Weight: 103,235 g/mol
MDL: MFCD00049640
CAS-Nummer: 12070-14-3
EINECS: 235-125-1
UN: 3178
ADR: 4.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR