3-Thiophensulfonylchlorid

MAYBCC13103CBEA 0 EUR
MAYBCC13103CB MAYBCC13103DA
3-Thiophensulfonylchlorid
3-Thiophensulfonylchlorid
Formel: C₄H₃ClO₂S₂
MDL: MFCD02682011
CAS-Nummer: 51175-71-4
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR