2-Fluorisonicotinsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL593885-250MGEA 94.2 EUR
SIAL593885-250MG
2-Fluorisonicotinsäure, Sigma-Aldrich®
2-Fluorisonicotinsäure
Formel: C₆H₄FNO₂
Molecular Weight: 141,10
MDL: MFCD02181194
CAS-Nummer: 402-65-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR