3-(2,4-Dimethoxyphenyl)propionsäure, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL575666-5GEA
150
EUR
SIAL575666-5G
3-(2,4-Dimethoxyphenyl)propionsäure, Sigma-Aldrich®
3-(2,4-Dimethoxyphenyl)propionsäure
Formel:
C₁₁H₁₄O₄ Molecular Weight: 210,23 g/mol |
MDL:
MFCD00598205 CAS-Nummer: 22174-29-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...