3-(2,3-Dimethoxyphenyl)propionsäure, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL568260-5GEA
50
EUR
SIAL568260-5G
3-(2,3-Dimethoxyphenyl)propionsäure, Sigma-Aldrich®
3-(2,3-Dimethoxyphenyl)propionsäure
Formel:
C₁₁H₁₄O₄ Molecular Weight: 210,23 |
MDL:
MFCD00029821 CAS-Nummer: 10538-48-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...