4'-tert-Butylacetanilid
Lieferant: Thermo Scientific
MAYBRF01502.50EA
0
EUR
MAYBRF01502.50
MAYBRF01502.1
MAYBRF01502.10
4'-tert-Butylacetanilid
4'-tert-Butylacetanilid
Formel:
C₁₂H₁₇NO |
MDL:
MFCD00043734 CAS-Nummer: 20330-45-4 EINECS: 000-000-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...