5-tert-Butyl-o-anisidin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL412627-10GEA 67.8 EUR
SIAL412627-10G
5-tert-Butyl-o-anisidin, Sigma-Aldrich®
5-tert-Butyl-o-anisidin
Formel: C₁₁H₁₇NO
Molecular Weight: 179,26
MDL: MFCD00026299
CAS-Nummer: 3535-88-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR