Ethylabietat

Lieferant: ABCR GMBH

ABCRAB135441.0025EA 0 EUR
ABCRAB135441.0025
Ethylabietat
Ethylabietat
Formel: C₂₂H₃₄O₂
Molecular Weight: 330,51 g/mol
MDL: MFCD00028860
CAS-Nummer: 631-71-0
EINECS: 211-166-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR