2,2'-Bithiophene-5-boronic acid, 96%

H52851.MDEA 81.3 EUR
ALFAH52851.MD
2,2'-Bithiophene-5-boronic acid, 96%
Purity: 96%
CAS No.: 132898-95-4
Molecular Formula: C8H7BO2S2
Formula Weight: 210.07
Melting Point: 163-165°
MDL No.: MFCD03086112
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR