5-Brom-2,1,3-benzothiadiazol

Lieferant: Thermo Scientific

MAYBCC09210.250EA 0 EUR
MAYBCC09210.250 MAYBCC09210DA
5-Brom-2,1,3-benzothiadiazol
5-Bromo-2,1,3-benzothiadiazole
Formel: C₆H₃BrN₂S
Molecular Weight: 215,0700 g/mol
MDL: MFCD00460091
CAS-Nummer: 1753-75-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR