2-Nitro-4-thiophencarbonsäure

MAYBDFP00046DAEA 0 EUR
MAYBDFP00046DA MAYBDFP00046CB
2-Nitro-4-thiophencarbonsäure
2-Nitro-4-thiophencarbonsäure
Formel: C₅H₃NO₄S
Molecular Weight: 173,15
MDL: MFCD00052582
CAS-Nummer: 40357-96-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR