3-Aminothiophen-2-carboxamid
Lieferant: Thermo Scientific
MAYBGK00439.250EA
0
EUR
MAYBGK00439.250
CHMPGK00439.0.1
3-Aminothiophen-2-carboxamid
3-Aminothiophen-2-carboxamid
Formel:
C₅H₆N₂OS Molecular Weight: 142,18 g/mol |
MDL:
MFCD00052593 CAS-Nummer: 147123-47-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...