4-Brom-2,5-dichlor-3-thiophensulfonylchlorid
Lieferant: Thermo Scientific
MAYBCD09555.10EA
0
EUR
MAYBCD09555.10
MAYBCD09555DA
4-Brom-2,5-dichlor-3-thiophensulfonylchlorid
4-Brom-2,5-dichlor-3-thiophensulfonylchlorid
Formel:
C₄BrCl₃O₂S₂ |
MDL:
MFCD00051664 CAS-Nummer: 166964-36-9 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...