4-Methoxypyridin-N-oxid Hydrat, Sigma-Aldrich®

Lieferant: Merck

SIAL65335-25G-FEA 230 EUR
SIAL65335-25G-F
4-Methoxypyridin-N-oxid Hydrat, Sigma-Aldrich®
4-Methoxypyridin-N-oxid Hydrat
Formel: C₆H₇NO₂·H₂O
Molecular Weight: 125,13 (anhy)
MDL: MFCD00149410
CAS-Nummer: 207511-18-0
EINECS: 214-367-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR