4-Chlorpyridin-N-oxid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL232408-5GEA 95.8 EUR
SIAL232408-5G
4-Chlorpyridin-N-oxid, Sigma-Aldrich®
4-Chlorpyridin-N-oxid
Formel: C₅H₄ClNO
MDL: MFCD00047425
CAS-Nummer: 1121-76-2
EINECS: 214-336-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR