Trieisenphosphid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL691593-5GEA 104 EUR
SIAL691593-5G
Trieisenphosphid, Sigma-Aldrich®
Triiron phosphide
Formel: Fe₃P
Molecular Weight: 198,51 g/mol
MDL: MFCD00799762
CAS-Nummer: 12023-53-9
EINECS: 234-682-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR