Dinickelphosphid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL372641-10GEA 232 EUR
SIAL372641-10G
Dinickelphosphid, Sigma-Aldrich®
Dinickel phosphide
Formel: Ni₂P
MDL: MFCD00016260
CAS-Nummer: 12035-64-2
EINECS: 234-828-0
UN: 3134
ADR: 4.3,I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR