Dimagnesiumsilicid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ200
SIAL343196-25GEA
123
EUR
SIAL343196-25G
Dimagnesiumsilicid, Sigma-Aldrich®
Dimagnesium silicide
Formel:
Mg₂Si Molecular Weight: 76,70 g/mol |
MDL:
MFCD00016202 CAS-Nummer: 22831-39-6 EINECS: 245-254-5 UN: 2624 ADR: 4.3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...