Calciumdisilicid

Lieferant: SILANE (PART OF CHEMPUR)

SILNSILS03575.50EA 0 EUR
SILNSILS03575.50
Calciumdisilicid
Calcium disilicide
Formel: CaSi₂
Molecular Weight: 96,25 g/mol
MDL: MFCD00015977
CAS-Nummer: 12013-56-8
EINECS: 234-588-7
UN: 1405
ADR: 4.3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR